Learn More
Description
Host Species: Mouse
Clone Q3-695
Isotype IgG1, κ
Species Reactivity Hu, Ms
Immunogen Human XBP-1S Recombinant Protein
Flow Cytometry, Intracellular Staining
Specifications
Specifications
| Antigen | XBP-1S |
| Classification | Monoclonal |
| Clone | Q3-695 |
| Conjugate | Alexa Fluor 647 |
| Host Species | Mouse |
| Immunogen | Human XBP-1S Recombinant Protein |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.