Learn More
XBP-1S Mouse, PE, Clone: Q3-695, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562642
Description
The Q3-695 monoclonal antibody specifically binds to XBP-1S, the spliced isoform of X-box binding protein 1 (XBP-1). XBP-1S is generated by cells that undergo an unfolded protein response, ie, cells responding to the accumulation of unfolded proteins in the endoplasmic reticulum. It plays important roles as a transcription factor in a number of important cellular processes including the regulation of MHC class II genes, differentiation of plasma cells, immunoglobulin secretion and hepatocyte growth.

Specifications
XBP-1S | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human XBP-1S Recombinant Protein | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
Q3-695 | |
XBP1; X-box binding protein 1; TREB5; Tax-responsive element-binding | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.