Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC7A11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA116775
Description
This antibody is predicted to react with mouse and canine based on 100% sequence homology. This antibody has 94% sequence homology with rat. Suggested positive control: human stomach protein, stomach tissue lysate. Immunogen sequence homology: orangutan (82%).
SLC7A11 (also known as XCT) is a member of a hetromeric, sodium-independent, anionic amino acid transport system. It is highly specific for cysteine and glutamate. SLC7A11 is a mediator of Kaposi sarcoma-associated herpes virus fusion and entry into cells. Increased expression of this protein in primary gliomas is associated with neuronal cell death.
Specifications
| SLC7A11 | |
| Polyclonal | |
| Unconjugated | |
| SLC7A11 | |
| 9930009M05Rik; AI451155; Amino acid transport system xc-; calcium channel blocker resistance protein CCBR1; CCBR1; cysteine/glutamate transporter; cystine/glutamate transporter; SLC7A11; sodium independent anionic amino acid transport system; solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11; solute carrier family 7 (cationic amino acid transporter, y+ system), member 11; solute carrier family 7 member 11; solute carrier family 7, (cationic amino acid transporter, y+ system) member 11; sut; xCT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23657, 26570, 310392 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9UPY5, Q9WTR6 | |
| SLC7A11 | |
| Synthetic peptide made to a region within the N-terminus of the mouse xCT protein (between residues 1-50). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction