Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XIAP Mouse, Unlabeled, Clone: 28, BD
Mouse Monoclonal Antibody
$286.00 - $566.00
Specifications
Antigen | XIAP |
---|---|
Clone | 28 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Apoptosis is a genetically programmed, selective process of cell death that occurs during normal cell differentiation and development of multicellular organisms. Viruses depend on the biosynthetic machinery of their host cell for the production of progeny and survival. Therefore, many viruses encode proteins that protect the cell from apoptosis. hILP ( h uman I AP- l ike p rotein) is a human homologue of the viral IAP ( I nhibitor of A poptosis P rotein). hILP is a widely expressed cytoplasmic protein of 497 amino acids with three BIR (Baculovirus IAP repeats) domains and a C-terminal RING finger domain. hILP-transfected cells are protected against the apoptotic effects of Sindbis virus infection and ICE (interleukin-1 β; converting enzyme) expression. This product is sold under license from Aegera Therapeutics, Inc.This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
XIAP | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
hILP |
28 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse | |
Human hILP/XIAP aa. 268-426 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title