Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XIAP Mouse, Unlabeled, Clone: 48, BD

Catalog No. BDB610762 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610762 50 μg
BDB610763 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610762 Supplier BD Biosciences Supplier No. 610762
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apoptosis is a genetically programmed, selective process of cell death that occurs during normal cell differentiation and development of multicellular organisms. Viruses depend on the biosynthetic machinery of their host cell for the production of progeny and survival. Therefore, many viruses encode proteins that protect the cell from apoptosis. hILP ( h uman I AP- l ike p rotein) is a human homologue of the viral IAP ( I nhibitor of A poptosis P rotein). hILP is a widely expressed cytoplasmic protein of 497 amino acids with three BIR (Baculovirus IAP repeats) domains and a C-terminal RING finger domain. hILP-transfected cells are protected against the apoptotic effects of Sindbis virus infection and ICE (interleukin-1 β; converting enzyme) expression. This product is sold under license from Aegera Therapeutics, Inc.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XIAP
Applications Western Blot
Classification Monoclonal
Clone 48
Concentration 250μg/mL
Conjugate Unconjugated
Description hILP
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human hILP aa. 268-426
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.