Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xin Mouse anti-Mouse, Unlabeled, Clone: 41, BD

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 611524

Catalog No. BDB611524


Description

Description

Normal cardiac morphogenesis requires a number of transcription factors, including Nkx2.5, MEF2C, eHand, and dHand. Both bone morphogenetic protein-(BMP) and Nkx2.5- dependent pathways have been implicated in intrinsic cardiogenesis. Xin, a BMP-induced protein, is expressed in embryonic and adult cardiac and skeletal muscle. It contains an N-terminal DNA binding domain, followed by a region of 16-amino acid repeating units, a nuclear localization signal, and a SH3-binding motif. In adult mice, Xin colocalizes with N-cadherin and connexin-43 in the intercalated discs of the heart, while in neonatal cardiomyocytes, Xin is found at cell-cell contacts and in the nucleus. In chick embryos, incubation with antisense oligonucleotides to cXin causes abnormal cardiac morphogenesis and an alteration in cardiac tube looping. In addition, BMP induces the expression of Nkx2.5, MEF2C, and cXin in chick embryos. This induction of Xin may occur downstream in this pathway, since both Nkx2.5 and MEF2C can act as Xin transactivators. Thus, Xin is thought to be involved in cell-cell junction dynamics and nuclear signaling during BMP- and Nkx2.5-dependent cardiac morphogenesis.

Host Species: Mouse
Clone: 41
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse Xin aa. 3-213
Formula Weight [Chemical]: 182kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Xin
Monoclonal
250μg/mL
Mouse
Affinity Purified
RUO
Primary
IgG1
Western Blot
41
Unconjugated
Mouse Xin aa. 3-213
50 μg
Cell Biology
Mouse
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.