Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

X-linked Kx blood group (McLeod syndrome), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006454 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-454 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-454 Supplier Abnova Corporation Supplier No. H00007504B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human XK protein.

This locus controls the synthesis of the Kell blood group 'precursor substance' (Kx). Mutations in this gene have been associated with McLeod syndrome, an X-linked, recessive disorder characterized by abnormalities in the neuromuscular and hematopoietic systems. The encoded protein has structural characteristics of prokaryotic and eukaryotic membrane transport proteins. [provided by RefSeq]

Sequence: MKFPASVLASVFLFVAETTAALSLSSTYRSGGDRMWQALTLLFSLLPCALVQLTLLFVHRDLSRDRPLVLLLHLLQLGPLFRCFEVFCIYFQSGNNEEPYVSITKKRQMPKNGLSEEIEKEVGQAEGKLITHRSAFSRASVIQAFLGSAPQLTLQLYISVMQQDVTVGRSLLMTISLLSIVYGALRCNILAIKIKYDEYEVKVKPLAYVCIFLWRSFEIATRVVVLVLFTSVLKTWVVVIILINFFSLFLYPWILFWCSGSPFPENIEKALSRVGTTIVLCFLTLLYTGINMFCWSAVQLKIDSPDLISKSHNWYQLLVYYMIRFIENAILLLLWYLFKTDIYMYVCAPLLVLQLLIGYCTAILFMLVFYQFFHPCKKLFSSSVSEGFQRWLRCFCWACRQQKPCEPIGKEDLQSSRDRDETPSSSKTSPEPGQFLNAEDLCSA

Specifications

Antigen X-linked Kx blood group (McLeod syndrome)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human XK protein.
Formulation No additive
Gene XK
Gene Accession No. BC036019.1
Gene Alias KX/X1k/XKR1
Gene Symbols XK
Host Species Mouse
Immunogen XK (AAH36019.1, 1 a.a. ∼ 444 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7504
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.