Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XKR4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535059
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
The specific function of the protein remains unknown.
Specifications
XKR4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q5GH76 | |
XKR4 | |
KLH conjugated synthetic peptide between 570-598 amino acids from the C-terminal region of human XKR4 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
XKR4 | |
XKR4; KIAA1889; XRG4; XK-related protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114786 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction