Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA527107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527107 Supplier Thermo Scientific Supplier No. PA527107

Rabbit Polyclonal Antibody

PA5-27108 targets beta Tubulin 2 in WB applications and shows reactivity with Drosophila and Human samples. The PA5-27108 immunogen is recombit fragment corresponding to a region within amino acids 1 and 356 of Human beta Tubulin 2.

The protein encoded by this gene functions together with DNA ligase IV and the DNA-dependent protein kinase in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. The non-homologous end-joining pathway is required both for normal development and for suppression of tumors. This gene functionally complements XR-1 Chinese hamster ovary cell mutant, which is impaired in DNA double-strand breaks produced by ionizing radiation and restriction enzymes. Alternative transcription initiation and alternative splicing generates several transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen XRCC4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene XRCC4
Gene Accession No. Q13426
Gene Alias DNA repair protein XRCC4, X-ray repair cross-complementing protein 4, X-ray repair, complementing defective, repair in Chinese hamster
Gene Symbols XRCC4
Host Species Rabbit
Immunogen Recombit fragment corresponding to a region within amino acids 1 and 109 of Human XRCC4
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7518
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.