Learn More
XRCC6, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human XRCC6 peptide using ARM Technology.
Supplier: Abnova Corporation H00002547K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The p70/p80 autoantigen is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
XRCC6 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
XRCC6 | |
XRCC6 | |
A synthetic peptide of human XRCC6 | |
RUO | |
2547 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
X-ray repair complementing defective repair in Chinese hamster cells 6 | |
XRCC6 | |
CTC75, CTCBF, G22P1, KU70, ML8, TLAA | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.