Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Y14 Mouse anti-Human, Rat, Mouse, Clone: 1F12, MilliporeSigma™

Catalog No. 041464MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-146-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-146-4MI Supplier MilliporeSigma Supplier No. 041464

Mouse Monoclonal Antibody

Use Anti-Y14 Antibody, clone 1F12 (Mouse Monoclonal Antibody) validated in western blot, immunoprecipitation, immunocytochemistry to detect Y14 also known as Binder of OVCA1-1, RNA binding motif protein 8A, Ribonucleoprotein RBM8A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Y14
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1F12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_005096.1
Gene Symbols RBM8A, BOV-1, BOV-1A, BOV-1B, BOV-1C, MDS014, RBM8A, RBM8B, Y14, ZNRP, ZRNP1
Host Species Mouse
Immunogen Purified human Y14.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.