Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Y14 Mouse anti-Human, Rat, Mouse, Clone: 4C4, MilliporeSigma™

Catalog No. 051511MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-151-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-151-1MI Supplier MilliporeSigma Supplier No. 051511
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Y14 Antibody, clone 4C4 is a Mouse Monoclonal Antibody for detection of Y14 also known as Binder of OVCA1-1, RNA binding motif protein 8A, Ribonucleoprotein RBM8A and has been validated in western blot, immunocytochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Y14
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4C4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_005096.1
Gene Symbols RBM8A, BOV-1, BOV-1A, BOV-1B, BOV-1C, MDS014, RBM8A, RBM8B, Y14, ZNRP, ZRNP1
Host Species Mouse
Immunogen Purified human Y14.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.