Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA519677
Description
This antibody is predicted to react with mouse and Xenopus laevis based on sequence homology.
This gene encodes the human ortholog of chicken YAP protein which binds to the SH3 domain of the Yes proto-oncogene product. This protein contains a WW domain that is found in various structural, regulatory and signaling molecules in yeast, nematode, and mammals, and may be involved in protein-protein interaction.
Specifications
YAP1 | |
Polyclonal | |
Unconjugated | |
P46937, P46938, Q2EJA0 | |
YAP1 | |
Synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Human YAP1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
YAP1 | |
YAP, YAP2, YAP65, YKI, 65 kDa Yes-associated protein; Yes-associated protein 1, 65kDa; yes-associated protein 2; yes-associated protein beta; yes-associated protein delta; yorkie homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10413, 22601, 363014 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction