Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yes Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB610376 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610376 150 μg
BDB610375 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610376 Supplier BD Biosciences Supplier No. 610376
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Yes is a member of the Src family of tyrosine kinases. The cellular gene was initially identified as a homologue of v-yes, the oncogene of avian sarcoma virus Y73. At least nine different Src family members have been identified. yes, fyn, and Src are widely expressed in a variety of cell types, while the remaining members are primarily expressed in hematopoietic cells. Members of this family have several common features: they have unique N-terminal domains, they attach to cellular membranes through a myristylated N-terminus and they have homologus SH2, SH3, and catalytic domains. The common expression patterns of Yes, fyn, and Src suggest some overlapping functions. All three of these kinases can physically associate with a number of cell-surface molecules which appear to increase their catalytic activity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Yes
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.