Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide, Mouse, Clone: 3G3, Abnova™

Catalog No. 89001052 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-052 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-052 Supplier Abnova Corporation Supplier No. H00007532M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant YWHAG.

This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the rat ortholog. It is induced by growth factors in human vascular smooth muscle cells, and is also highly expressed in skeletal and heart muscles, suggesting an important role for this protein in muscle tissue. It has been shown to interact with RAF1 and protein kinase C, proteins involved in various signal transduction pathways. [provided by RefSeq

Sequence: EQKTSADGNEKKIEMVRAYREKIEKELEAVCQDVLSLLDNYLIKNCSETQYESKVFYLKMKGDYYRYLAEVATGEKRATVVESSEKAYSEAHEISKEHMQ

Specifications

Antigen tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide
Applications ELISA
Classification Monoclonal
Clone 3G3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant YWHAG.
Formulation PBS with no preservative; pH 7.4
Gene YWHAG
Gene Accession No. NM_012479
Gene Alias 14-3-3GAMMA
Gene Symbols YWHAG
Host Species Mouse
Immunogen YWHAG (NP_036611, 67 a.a. ∼ 166 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7532
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.