Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 Mouse anti-Human, Mouse, Clone: 1E7.2, MilliporeSigma™

Catalog No. 05776MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-776-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-776-MI Supplier MilliporeSigma Supplier No. 05776
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-ZAP-70 Antibody, clone 1E7.2 is an antibody against ZAP-70 for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAP-70
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1E7.2
Conjugate Unconjugated
Formulation of 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_001079
Gene Symbols ZAP70, ZAP-70, TZK, STD, SRK
Host Species Mouse
Immunogen GST fusion protein between the second SH2 domain and precedes the kinase domain corresponding to residues 282-307 of human ZAP-70
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.