Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 Mouse anti-Human, Clone: 2F3.2, MilliporeSigma™

Catalog No. 05253MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-253-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-253-MI Supplier MilliporeSigma Supplier No. 05253
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-ZAP-70 Antibody, clone 2F3.2 is validated for use in flow cytometry, immunohistochemistry, immunoprecipitation, western blotting for the detection of ZAP-70.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAP-70
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2F3.2
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative.
Gene Accession No. NM_001079
Gene Symbols ZAP70, ZAP-70, TZK, STD, SRK
Host Species Mouse
Immunogen GST-fusion to tandem SH2 domains of human ZAP-70 corresponding to residues 1-254
Purification Method Protein G purfied
Quantity 250 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.