Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 Mouse anti-Human, Clone: 2F3.2, MilliporeSigma™

Catalog No. 05253MGMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-253-MGMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-253-MGMI Supplier MilliporeSigma Supplier No. 05253MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect ZAP-70 using this Anti-ZAP-70 Antibody, clone 2F3.2 validated for use in flow cytometry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAP-70
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2F3.2
Conjugate Unconjugated
Formulation PBS
Gene Accession No. NM_001079
Gene Symbols ZAP70, ZAP-70, TZK, STD, SRK
Host Species Mouse
Immunogen GST-fusion to tandem SH2 domains of human ZAP-70 corresponding to residues 1-254
Purification Method Protein G chromatography
Quantity 1 mg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.