Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 Mouse anti-Human, PE, Clone: 1E7.2, BD

Catalog No. BDB344635 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB344635 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB344635 Supplier BD Biosciences Supplier No. 344635
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti–ZAP-70, clone 1E7.2, is generated from the fusion of AG8.553 myeloma cells with spleen cells from BALB/c mice immunized with a ZAP-70 peptide 282-307 conjugated to KLH.

Anti–ZAP-70 is a 70-kilodalton (kd) protein kinase, a member of the Syk family of protein tyrosine kinases that is associated with the zeta chain of the CD3/TCR complex. Anti–ZAP-70 plays a crucial role in T-cell signal transduction and thymocyte development.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAP-70
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1E7.2
Concentration 12.5μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen ZAP-70 peptide 282-307 conjugated to KLH
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.