Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zap70 (pY292) Mouse anti-Human, Alexa Fluor 647, Clone: J34-602, BD

Catalog No. BDB558515 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558515 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558515 Supplier BD Biosciences Supplier No. 558515

Mouse Monoclonal Antibody

The 70kDa ζchain-associated protein (ZAP70) is a Syk-family protein tyrosine kinase (PTK) that associates with the ζ subunit of the T cell antigen receptor (TCR) and undergoes tyrosine phosphorylation following TCR stimulation. ZAP70 contains two SH2-like domains with the PTK domain located at the C-terminus. TCR-mediated Lck activity leads to phosphorylation of ZAP70 on Tyrosine 493 in the regulatory loop of the PTK domain leading to upregulation of ZAP70 kinase activity. Tyrosine 292 (Y292), in the linker region between the SH2 and PTK domains, is autophosphorylated by the activated PTK domain. By binding with c-Cbl, the phosphorylated Y292 can down-regulate TCR signaling. The J34-602 antibody recognizes ZAP70 phosphorylated at Y292.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen Zap70 (pY292)
Applications Flow Cytometry
Classification Monoclonal
Clone J34-602
Conjugate Alexa Fluor 647
Description ZAP-70 (pY292)
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human ZAP70
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.