Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBP1/DLM-1/DAI Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176854
Description
ZBP1/DLM-1/DAI Polyclonal specifically detects ZBP1/DLM-1/DAI in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Knockdown Validated.Specifications
ZBP1/DLM-1/DAI | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
C20orf183, chromosome 20 open reading frame 183, DAI, DLM1, DLM-1, DLM1dJ718J7.3, DNA-dependent activator of interferon regulatory factors, DNA-dependent activator of IRFs, Tumor stroma and activated macrophage protein DLM-1, ZBP1 Z-DNA binding protein 1, Z-DNA binding protein 1, Z-DNA-binding protein 1 | |
Rabbit | |
42 kDa | |
0.1 mg | |
Primary | |
Multiple isoforms of ZBP1 are known to exist. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
1 mg/mL | |
Western Blot 0.5-2 ug/ml, Simple Western reported by internal validation, ELISA 1:100-1:2000, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 2.5 ug/ml, Knockdown Validated | |
EAW75510 | |
ZBP1 | |
Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human ZBP1. The immunogen is located within the last 50 amino acids of ZBP1. Amino Acid Squence: TPSHSKLTPKLETMTLGN | |
Affinity Purified | |
RUO | |
81030 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction