Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger and BTB domain containing 7B, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007579 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-579 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-579 Supplier Abnova Corporation Supplier No. H00051043B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ZBTB7B protein.

ZFP67 is an early growth response gene that encodes a zinc finger-containing transcription factor that binds to the promoter regions of type I collagen genes (e.g., COL1A1; MIM 120150) and has a role in development.[supplied by OMIM

Sequence: MGSPEDDLIGIPFPDHSSELLSCLNEQRQLGHLCDLTIRTQGLEYRTHRAVLAACSHYFKKLFTEGGGGAVMGAGGSGTATGGAGAGVCELDFVGPEALGALLEFAYTATLTTSSANMPAVLQAARLLEIPCVIAACMEILQGSGLEAPSPDEDDCERARQYLEAFATATASGVPNGEDSPPQVPLPPPPPPPPRPVARRSRKPRKAFLQTKGARANHLVPEVPTVPAHPLTYEEEEVAGRVGSSGGSGPGDSYSPPTGTASPPEGPQSYEPYEGEEEEEELVYPPAYGLAQGGGPPLSPEELGSDEDAIDPDLMAYLSSLHQDNLAPGLDSQDKLVRKRRSQMPQECPVCHKIIHGAGKLPRHMRTHTGEKPFACEVCGVRFTRNDKLKIHMRKHTGERPYSCPHCPARFLHSYDLKNHMHLHTGDRPYECHLCHKAFAKEDHLQRHLKGQNCLEVRTRRRRKDDAPPHYPPPSTAAASPAGLDLSNGHLDTFRLSLARFWEQSAPTGPPVSTPGPPDDDEEEGAPTTPQAEGAMESS

Specifications

Antigen zinc finger and BTB domain containing 7B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ZBTB7B protein.
Formulation No additive
Gene ZBTB7B
Gene Accession No. BC012070.1
Gene Alias DKFZp686G01254/THPOK/ZBTB15/ZFP67/ZNF857B/c-Krox/hcKrox
Gene Symbols ZBTB7B
Host Species Mouse
Immunogen ZBTB7B (AAH12070.1, 1 a.a. ∼ 539 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 51043
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.