Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zDHHC8 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB15242MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15242MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15242MI Supplier MilliporeSigma Supplier No. AB15242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-zDHHC8 Antibody is an antibody against zDHHC8 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen zDHHC8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in PBS with 0.05% NaN3
Gene Symbols DHHC-8; EC 2.3.1.- ; KIAA1292; ZDHHCL1; ZNF378
Host Species Rabbit
Immunogen Syntheic linear peptide.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) Q9ULC8
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Handling Recommendations: Prior to removing the cap gently centrifuge the vial and gently mix the solution.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.