Learn More
ZFHX1B, Mouse, Clone: 2D9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ZFHX1B.
Supplier: Abnova Corporation H00009839M01
Description
The SMADIP1 gene (also known as SIP1) is a member of the delta-EF1 (ZEB1; MIM 189909)/Zfh1 family of 2-handed zinc finger/homeodomain proteins. SMADIP1 interacts with receptor-mediated, activated full-length SMADs (see MIM 605568) (Verschueren et al., 1999 [PubMed 10400677]).[supplied by OMIM
Sequence: SITPQGYSDSEERESMPRDGESEKEHEKEGEDGYGKLGRQDGDEEFEEEEEESENKSMDTDPETIRDEEETGDHSMDDSSEDGKMETKSDHEEDNMEDGMSpecifications
ZFHX1B, | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_014795.2 | |
ZEB2 | |
ZFHX1B (NP_055610.1, 1115 a.a. ∼ 1214 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2D9 | |
Mouse monoclonal antibody raised against a partial recombinant ZFHX1B. | |
ZEB2 | |
KIAA0569/SIP-1/SIP1/SMADIP1/ZFHX1B | |
Mouse | |
Affinity chromatography | |
RUO | |
9839 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.