Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFHX1B, Mouse, Clone: 4G8, Abnova™

Catalog No. 89002762 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-762 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-762 Supplier Abnova Corporation Supplier No. H00009839M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ZFHX1B.

The SMADIP1 gene (also known as SIP1) is a member of the delta-EF1 (ZEB1; MIM 189909)/Zfh1 family of 2-handed zinc finger/homeodomain proteins. SMADIP1 interacts with receptor-mediated, activated full-length SMADs (see MIM 605568) (Verschueren et al., 1999 [PubMed 10400677]).[supplied by OMIM

Sequence: SITPQGYSDSEERESMPRDGESEKEHEKEGEDGYGKLGRQDGDEEFEEEEEESENKSMDTDPETIRDEEETGDHSMDDSSEDGKMETKSDHEEDNMEDGM

Specifications

Antigen ZFHX1B,
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4G8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ZFHX1B.
Formulation PBS with no preservative; pH 7.4
Gene ZEB2
Gene Accession No. NM_014795.2
Gene Alias KIAA0569/SIP-1/SIP1/SMADIP1/ZFHX1B
Gene Symbols ZEB2
Host Species Mouse
Immunogen ZFHX1B (NP_055610.1, 1115 a.a. ∼ 1214 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9839
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.