Learn More
Description
This gene encodes a transcription factor with multiple homeodomains and zinc finger motifs, and regulates myogenic and neuronal differentiation. The encoded protein suppresses expression of the alpha-fetoprotein gene by binding to an AT-rich enhancer motif. The protein has also been shown to negatively regulate c-Myb, and transactivate the cell cycle inhibitor cyclin-dependent kinase inhibitor 1A (also known as p21CIP1). This gene is reported to function as a tumor suppressor in several cancers, and sequence variants of this gene are also associated with atrial fibrillation. Multiple transcript variants expressed from alternate promoters and encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | ZFHX3 |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 3B1 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant ZFHX3. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZFHX3 |
| Gene Accession No. | NM_006885 |
| Gene Alias | ATBF1/ATBT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.