Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP36L1, Mouse, Clone: 1A3, Abnova™

Catalog No. 89016366 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-366 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-366 Supplier Abnova Corporation Supplier No. H00000677M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ZFP36L1.

This gene is a member of the TIS11 family of early response genes. Family members are induced by various agonists such as the phorbol ester TPA and the polypeptide mitogen EGF. The gene is well conserved across species and has a promoter that contains motifs seen in other early-response genes. The encoded protein contains a distinguishing putative zinc finger domain with a repeating cys-his motif. This putative nuclear transcription factor most likely functions in regulating the response to growth factors. [provided by RefSeq

Sequence: MTTTLVSATIFDLSEVLCKGNKMLNYSAPSAGGCLLDRKAVGTPAGGGFPRRHSVTLPSSKFHQNQLLSSLKGEPAPALSSRDSRFRDRSFSEGGERLLPTQKQPGGG

Specifications

Antigen ZFP36L1
Applications ELISA, KnockDown, Western Blot
Classification Monoclonal
Clone 1A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ZFP36L1.
Formulation PBS with no preservative; pH 7.4
Gene ZFP36L1
Gene Accession No. NM_004926
Gene Alias BRF1/Berg36/ERF-1/ERF1/RNF162B/TIS11B/cMG1
Gene Symbols ZFP36L1
Host Species Mouse
Immunogen ZFP36L1 (NP_004917, 1 a.a. ∼ 108 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 677
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.