Learn More
Description
Includes
Up to three rabbit IgG clones of 1mg each will be delivered to customer.
The ZFYVE16 gene encodes endofin, an endosomal protein implicated in regulating membrane trafficking. It is characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain positioned in the middle of the molecule (Seet et al., 2004 [PubMed 14613930]).[supplied by OMIM
Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
Specifications
| Antigen | ZFYVE16 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Concentration | 1mg |
| Conjugate | Unconjugated |
| Description | zinc finger, FYVE domain containing 16 |
| Formulation | In 1X PBS, pH 7.4 |
| Gene | ZFYVE16 |
| Gene Accession No. | ZFYVE16 |
| Gene Alias | DKFZp686E13162, ENDOFIN, KIAA0305 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.