Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZFYVE28 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524077
Description
Negative regulator of epidermal growth factor receptor (EGFR) signaling. Acts by promoting EGFR degradation in endosomes when not monoubiquitinated.
Specifications
| ZFYVE28 | |
| Polyclonal | |
| ZFYVE28 | |
| LST2, LYST2, hLst2, lateral signaling target protein 2 homolog, zinc finger FYVE domain-containing protein 28 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 231125, 57732 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q9HCC9, Q6ZPK7 | |
| ZFYVE28 | |
| KLH conjugated synthetic peptide between 604-633 amino acids from the C-terminal region of human ZFYVE28 | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction