Learn More
Zic family member 1 (odd-paired homolog, Drosophila), Mouse, Clone: 2D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ZIC1.
Supplier: Abnova Corporation H00007545M02
Description
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene. [provided by RefSeq
Sequence: LLDAGPQYPAIGVTTFGASRHHSAGDVAERDVGLGINPFADGMGAFKLNPSSHELASAGQTAFTSQAPGYAAAAALGHHHHPGHVGSYSSAAFNSpecifications
Zic family member 1 (odd-paired homolog, Drosophila) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003412 | |
ZIC1 | |
ZIC1 (NP_003403, 2 a.a. ∼ 95 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Signal Transduction | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
2D3 | |
Mouse monoclonal antibody raised against a partial recombinant ZIC1. | |
ZIC1 | |
ZIC/ZNF201 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
7545 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.