Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zic3 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB15466MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15466MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15466MI Supplier MilliporeSigma Supplier No. AB15466

Rabbit Polyclonal Antibody

This Anti-Zic3 Antibody is validated for use in western blotting for the detection of Zic3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zic3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS with 0.1% sodium azide.
Gene Symbols ZIC3; HTX1; ZNF203; HTX
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_003413.2
Target Species Human
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.