Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 136, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018570 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-570 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-570 Supplier Abnova Corporation Supplier No. H00007695D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ZNF136 protein.

Sequence: MDSVAFEDVDVNFTQEEWALLDPSQKNLYRDVMWETMRNLASIGKKWKDQNIKDHYKHRGRNLRSHMLERLYQTKDGSQRGGIFSQFANQNLSKKIPGVKLCESIVYGEVSMGQSSLNRHIKDHSGHEPKEYQEYGEKPDTRNQCWKPFSSHHSFRTHEIIHTGEKLYDCKECGKTFFSLKRIRRHIITHSGYTPYKCKVCGKAFDYPSRFRTHERSHTGEKPYECQECGKAFTCITSVRRHMIKHTGDGPYKCKVCGKPFHSLSSFQVHERIHTGEKPFKCKQCGKAFSCSPTLRIHERTHTGEKPYECKQCGKAFSYLPSLRLHERIHTGEKPFVCKQCGKAFRSASTFQIHERTHTGEKPYECKECGEAFSCIPSMRRHMIKHTGEGPYKCKVCGKPFHSLSPFRIHERTHTGEKPYVCKHCGKAFVSSTSIRIHERTHTGEKPYECKQCGKAFSYLNSFRTHEMIHTGEKPFECKRCGKAFRSSSSFRLHERTHTGQKPYHCKECGKAYSCRASFQRHMLTHAEDGPPYKCMWESL

Specifications

Antigen zinc finger protein 136
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ZNF136 protein.
Formulation PBS with no preservative; pH 7.4
Gene ZNF136
Gene Accession No. NM_003437.2
Gene Alias pHZ-20
Gene Symbols ZNF136
Host Species Rabbit
Immunogen ZNF136 (NP_003428.1, 1 a.a. ∼ 540 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 7695
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.