Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
zinc finger protein 174 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZNF174 protein.
Supplier: Abnova Corporation H00007727B01
Description
Sequence: MAAKMEITLSSNTEASSKQERHIIAKLEEKRGPPLQKNCPDPELCRQSFRRFCYQEVSGPQEALSQLRQLCRQWLQPELHTKEQILELLVMEQFLTILPPEIQARVRHRCPMSSKEIVTLVEDFHRASKKPKQWVAVCMQGQKVLLEKTGSQLGEQELPDFQPQTPRRDLRESSPAEPSQAGAYDRLSPHHWEKSPLLQEPTPKLAGTELLIEKTDPNMATDELPCKLWLSFIASpecifications
zinc finger protein 174 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ZNF174 protein. | |
ZNF174 | |
ZSCAN8 | |
Mouse | |
50 μL | |
Yes | |
7727 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC000876 | |
ZNF174 | |
ZNF174 (AAH00876, 1 a.a. ∼ 234 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction