Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 207, Mouse, Clone: 6D7, Abnova™

Catalog No. 89001063 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-063 Supplier Abnova Corporation Supplier No. H00007756M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ZNF207.

Sequence: MGRKKKKQLKPWCWYCNRDFDDEKILIQHQKAKHFKCHICHKKLYTGPGLAIHCMQVHKETIDAVPNAIPGRTDIELEIYGMEGIPEKDMDERRRLLEQKTQESQKKKQQ

Specifications

Antigen zinc finger protein 207
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 6D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ZNF207.
Formulation PBS with no preservative; pH 7.4
Gene ZNF207
Gene Accession No. NM_003457
Gene Alias DKFZp761N202
Gene Symbols ZNF207
Host Species Mouse
Immunogen ZNF207 (NP_003448, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7756
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.