Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
zinc finger protein 25, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZNF25 protein.
Supplier: Abnova Corporation H00219749B01
Description
Sequence: MNKFQGPVTLKDVIVEFTKEEWKLLTPAQRTLYKDVMLENYSHLVSVGYHVNKPNAVFKLKQGKEPWILEVEFPHRGFPEDLWSIHDLEARYQESQAGNSRNGELTKHQKTHTTEKACECKECGKFFCQKSALIVHQHTHSKGKSYDCDKCGKSFSKNEDLIRHQKIHTRDKTYECKECKKIFYHLSSLSRHLRTHAGEKPYECNQCEKSFYQKPHLTEHQKTHTGEKPFECTECGKFFYVKAYLMVHQKTHTGEKPYECKECGKAFSQKSHLTVHQRMHTGEKPYKCKECGKFFSRNSHLKTHQRSHTGEKPYECKECRKCFYQKSALTVHQRTHTGEKPFECNKCGKTFYYKSDLTKHQRKHTGEKPYECTECGKSFAVNSVLRLHQRTHTGEKPYACKECGKSFSQKSHFIIHQRKHTGEKPYECQECGETFIQKSQLTAHQKTHTKKRNAEKSpecifications
zinc finger protein 25 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ZNF25 protein. | |
ZNF25 | |
DKFZp564C206/FLJ31890/KOX19/Zfp9 | |
Mouse | |
50 μL | |
Yes | |
219749 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
NM_145011.2 | |
ZNF25 | |
ZNF25 (NP_659448.1, 1 a.a. ∼ 456 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction