Learn More
zinc finger protein 274, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ZNF274 protein.
Supplier: Abnova Corporation H00010782D01P
Description
This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals. [provided by RefSeq
Sequence: MASRLPTAWSCEPVTFEDVTLGFTPEEWGLLDLKQKSLYREVMLENYRNLVSVEHQLSKPDVVSQLEEAEDFWPVERGIPQDTIPEYPELQLDPKLDPLPAESPLMNIEVVEVLTLNQEVAGPRNAQIQALYAEDGSLSADAPSEQVQQQGKHPGDPEAARQRFRQFRYKDMTGPREALDQLRELCHQWLQPKARSKEQILELLVLEQFLGALPVKLRTWVESQHPENCQEVVALVEGVTWMSEEEVLPAGQPAEGTTCCLEVTAQQEEKQEDAAICPVTVLPEEPVTFQDVAVDFSREEWGLLGPTQRTEYRDVMLETFGHLVSVGWETTLENKELAPNSDIPEEEPAPSLKVQESSRDCALSSTLEDTLQGGVQEVQDTVLKQMESAQEKDLPQKKHFDNRESQANSGALDTNQVSLQKIDNPESQANSGALDTNQVLLHKIPPRKRLRKRDSQVKSMKHNSRVKIHQKSCERQKAKEGNGCRKTFSRSTKQITFIRIHKGSQVCRCSECGKIFRNPRYFSVHKKIHTGERPYVCQDCGKGFVQSSSLTQHQRVHSGERPFECQECGRTFNDRSAISQHLRTHTGAKPYKCQDCGKAFRQSSHLIRHQRTHTGERPYACNKCGKAFTQSSHLIGHQRTHNRTKRKKKQPTSSpecifications
zinc finger protein 274 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human ZNF274 protein. | |
ZNF274 | |
DKFZp686K08243/FLJ37843/HFB101/ZF2/ZKSCAN19 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_133502.1 | |
ZNF274 | |
ZNF274 (NP_598009.1, 1 a.a. ∼ 653 a.a) full-length human protein. | |
100 μg | |
Transcription Regulation | |
10782 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.