Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
zinc finger and SCAN domain containing 20, Mouse, Clone: 3G7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ZNF31.
Supplier: Abnova Corporation H00007579M02
Description
Sequence: MAMALELQAQASPQPEPEELLIVKLEEDSWGSESKLWEKDRGSVSGPEASRQRFRQFQYRDAAGPHEAFSQLWALCCRWLRPEIRLKEQILELLVLEQFLTILPREVQTWVQARHPESGEEAVALVEDWHRETRTAGQSGLELHTEETRPLKTGEEAQSFQLQPVDPWPEGQSQKKGVKNTCPDLPNHLNAEVAPQPLKESGVPVSKPSNTSEKEQGPEFWGLSLINSGKRSTADYSLDNEPAQALTWRDSRAWEEQYQWDVEDMKVSGVHWGYEETKTFLAILSESPFSEKLRTCHQNRQVYRAIAEQLRARGFLRTLEQCRYRVKNLLRNYRKAKSSHPPGTCPFYEELEALVRARTAIRATDGPGEAVALPRLGDSDAEMDEQEEGGWDPEEMAEDCNGAGLVNVESTQGPRIAGAPALFQSRIGKNMGVSpecifications
| zinc finger and SCAN domain containing 20 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC011404 | |
| ZSCAN20 | |
| ZNF31 (AAH11404, 1 a.a. ∼ 433 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 7579 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3G7 | |
| Mouse monoclonal antibody raised against a full length recombinant ZNF31. | |
| ZSCAN20 | |
| KOX29/ZFP-31/ZNF31/ZNF360 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction