Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF350, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017141 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-141 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-141 Supplier Abnova Corporation Supplier No. H00059348B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ZNF350 protein.

Sequence: MIQAQESITLEDVAVDFTWEEWQLLGAAQKDLYRDVMLENYSNLVAVGYQASKPDALFKLEQGEQLWTIEDGIHSGACSDIWKVDHVLERLQSESLVNRRKPCHEHDAFENIVHCSKSQFLLGQNHDIFDLRGKSLKSNLTLVNQSKGYEIKNSVEFTGNGDSFLHANHERLHTAIKFPASQKLISTKSQFISPKHQKTRKLEKHHVCSECGKAFIKKSWLTDHQVMHTGEKPHRCSLCEKAFSRKFMLTEHQRTHTGEKPYECPECGKAFLKKSRLNIHQKTHTGEKPYICSECGKGFIQKGNLIVHQRIHTGEKPYICNECGKGFIQKTCLIAHQRFHTGKTPFVCSECGKSCSQKSGLIKHQRIHTGEKPFECSECGKAFSTKQKLIVHQRTHTGERPYGCNECGKAFAYMSCLVKHKRIHTREKQEAAKVENPPAERHSSLHTSDVMQEKNSANGATTQVPSVAPQTSLNISGLLANRNVVLVGQPVVRCAASGDNRGFAQDRNLVNAVNVVVPSVINYVLFYVTENP

Specifications

Antigen ZNF350
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ZNF350 protein.
Formulation PBS with no preservative; pH 7.4
Gene ZNF350
Gene Accession No. NM_021632.3
Gene Alias ZBRK1/ZFQR
Gene Symbols ZNF350
Host Species Mouse
Immunogen ZNF350 (NP_067645.3, 1 a.a. ∼ 532 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59348
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.