Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 394, Mouse, Clone: 1F8, Abnova™

Catalog No. 89001368 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-368 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-368 Supplier Abnova Corporation Supplier No. H00084124M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ZNF394.

Sequence: MNSSLTAQRRGSDAELGPWVMAARSKDAAPSQRDGLLPVKVEEDSPGSWEPNYPAASPDPETSRLHFRQLRYQEVAGPEEALSRLRELCRRWLRPELLSKEQILELLVLEQFLTILPEELQAWVREHCPESGEEAVAVVRALQRALDGTSSQGMVTFEDTAVSLTWEEWERLDPARRDFCRESAQKDSGSTVPPSLESRVENKELIPMQQILEEAEPQGQLQEAFQGKRPLFSKCGSTHEDRVEKQSGDPLPLKLENSPEAEGLNSISDVNKNGSIEGEDSKNNELQNSARCSNLVLCQHIPKAERPTDSEEHGNKCKQSFHMVTWHVLKPHKSDSGDSFHHSSLFETQRQLHEERPYKCGNCGKSFKQRSDLFRHQRIHTGEKPYGCQECGKSFSQSAALTKHQRTHTGEKPYTCLKCGERFRQNSHLNRHQSTHSRDKHFKCEECGETCHISNLFRHQRLHKGERPYKCEECEKSFKQRSDLFKHHRIHTGEKPYGCSVCGKRFNQSATLIKHQRIHTGEKPYKCLECGERFRQSTHLIRHQRIHQNKVLSAGRGGSRL

Specifications

Antigen zinc finger protein 394
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant ZNF394.
Formulation PBS with no preservative; pH 7.4
Gene ZNF394
Gene Accession No. BC025241
Gene Alias FLJ12298/ZKSCAN14
Gene Symbols ZNF394
Host Species Mouse
Immunogen ZNF394 (AAH25241, 1 a.a. ∼ 561 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84124
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.