Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

zinc finger protein 394, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008314 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-314 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-314 Supplier Abnova Corporation Supplier No. H00084124B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ZNF394 protein.

Sequence: MNSSLTAQRRGSDAELGPWVMAARSKDAAPSQRDGLLPVKVEEDSPGSWEPNYPAASPDPETSRLHFRQLRYQEVAGPEEALSRLRELCRRWLRPELLSKEQILELLVLEQFLTILPEELQAWVREHCPESGEEAVAVVRALQRALDGTSSQGMVTFEDTAVSLTWEEWERLDPARRDFCRESAQKDSGSTVPPSLESRVENKELIPMQQILEEAEPQGQLQEAFQGKRPLFSKCGSTHEDRVEKQSGDPLPLKLENSPEAEGLNSISDVNKNGSIEGEDSKNNELQNSARCSNLVLCQHIPKAERPTDSEEHGNKCKQSFHMVTWHVLKPHKSDSGDSFHHSSLFETQRQLHEERPYKCGNCGKSFKQRSDLFRHQRIHTGEKPYGCQECGKSFSQSAALTKHQRTHTGEKPYTCLKCGERFRQNSHLNRHQSTHSRDKHFKCEECGETCHISNLFRHQRLHKGERPYKCEECEKSFKQRSDLFKHHRIHTGEKPYGCSVCGKRFNQSATLIKHQRIHTGEKPYKCLECGERFRQSTHLIRHQRIHQNKVLSAGRGGSRL

Specifications

Antigen zinc finger protein 394
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ZNF394 protein.
Formulation No additive
Gene ZNF394
Gene Accession No. NM_032164.2
Gene Alias FLJ12298/ZKSCAN14
Gene Symbols ZNF394
Host Species Mouse
Immunogen ZNF394 (NP_115540.2, 1 a.a. ∼ 561 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84124
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.