Learn More
zinc finger protein 41, Mouse, Clone: 4D6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ZNF41.
Supplier: Abnova Corporation H00007592M02
Description
This gene product is a likely zinc finger family transcription factor. It contains KRAB-A and KRAB-B domains that act as transcriptional repressors in related proteins, and multiple zinc finger DNA binding motifs and finger linking regions characteristic of the Kruppel family. This gene is part of a gene cluster on chromosome Xp11.23. Several alternatively spliced transcript variants have been described, however, the full-length nature of only some of them is known. [provided by RefSeq
Sequence: GNNFPHSPSSTKNENAKTGANSCEHDHYEKHLSHKQAPTHHQKIHPEEKLYVCTECVMGFTQKSHLFEHQRIHAGEKSRECDKSNKVFPQKPQVDVHPSV*Specifications
| zinc finger protein 41 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_007130 | |
| ZNF41 | |
| ZNF41 (NP_009061, 221 a.a. ∼ 321 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4D6 | |
| Mouse monoclonal antibody raised against a partial recombinant ZNF41. | |
| ZNF41 | |
| MGC8941/MRX89 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 7592 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.