Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF483 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535212
Description
ZNF483 may be involved in transcriptional regulation.
Specifications
| ZNF483 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q8TF39 | |
| ZNF483 | |
| KLH conjugated synthetic peptide between 281-309 amino acids from the central region of human ZNF483 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ZNF483 | |
| ZNF483; KIAA1962; ZKSCAN16; Zinc finger protein 483; Zinc finger protein with KRAB and SCAN domains 16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158399 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction