Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
zinc finger protein 496, Mouse, Clone: 4B1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ZNF496.
Supplier: Abnova Corporation H00084838M02
Description
Sequence: HLQPDRLQPVEKREQAASEDADKGPKEPLENGKAKLSFQCCECGKAFQRHDHLARHRSHFHLKDKARPFQCRYCVKSFTQNYDLLRHERLHMKRRSKQALNSpecifications
zinc finger protein 496 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_032752 | |
ZNF496 | |
ZNF496 (NP_116141, 485 a.a. ∼ 585 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
84838 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Immunofluorescence, Western Blot | |
4B1 | |
Mouse monoclonal antibody raised against a partial recombinant ZNF496. | |
ZNF496 | |
MGC15548/NIZP1/ZKSCAN17 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction