Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF639 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA531718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531718 Supplier Thermo Scientific Supplier No. PA531718

Rabbit Polyclonal Antibody

PA5-31718 targets ZNF639 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-31718 immunogen is recombit fragment corresponding to a region within amino acids 281 and 485 of Human ZNF639. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF639
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Gene ZNF639
Gene Accession No. Q9UID6
Gene Alias 6230400O18Rik, ANC-2H01, ANC_2H01, ZASC1, zinc finger amplified in esophageal squamous cell carcinomas 1, zinc finger protein ANC_2H01, zinc finger protein ZASC1
Gene Symbols ZNF639
Host Species Rabbit
Immunogen Recombit fragment corresponding to a region within amino acids 281 and 485 of Human ZNF639
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.