Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF683 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA130046

Catalog No. PIPA130046


Only null left
Add to Cart

Description

Description

Recommended positive controls: HepG2. Predicted reactivity: Human: 100%. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene, ZNF683, which is located on chromosome 1, is predicted to encode a zinc finger protein, currently with unknown fucntion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF683
Polyclonal
Unconjugated
ZNF683
Hobit; homolog of Blimp-1 in T cells; Homolog of Blimp-1 in T-cell; hypothetical protein MGC33414; MGC33414; RP11-569G9.6; Tissue-resident T-cell transcription regulator protein ZNF683; Zinc finger protein 683; ZNF683
Rabbit
Protein A
RUO
257101
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8IZ20
ZNF683
A synthetic peptide corresponding to a N-terminal region of Human ZNF683.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.