Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF9 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524717
Description
This antibody is predicted to react with bovine, chicken, mouse and rat based on sequence homology.
ZNF9 encodes a nucleic-acid binding protein with seven zinc-finger domains. The protein has a preference for binding single stranded DNA and RNA. The protein functions in cap-independent translation of ornithine decarboxylase mRNA, and may also function in sterol-mediated transcriptional regulation. A CCTG expansion in the first intron of this gene results in myotonic dystrophy type 2.
Specifications
ZNF9 | |
Polyclonal | |
CNBP | |
CNBP1, DM2, PROMM, RNF163, ZCCHC22, ZNF9, cellular nucleic acid binding protein, cellular nucleic acid-binding protein, erythroid differentiation-related, sterol regulatory element-binding protein, zinc finger protein 273, zinc finger protein 9 (a cellular retroviral nucleic acid binding protein) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12785, 7555 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P62633, P53996 | |
CNBP | |
KLH conjugated synthetic peptide between 67-96 amino acids from the central region of human ZNF9 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction