Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF98 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524025
Description
ZNF98 Polyclonal Antibody for Western Blot, IHC (P), Flow
May be involved in transcriptional regulation.
Specifications
| ZNF98 | |
| Polyclonal | |
| Unconjugated | |
| ZNF98 | |
| F7175; zinc finger protein 739; Zinc finger protein 98; zinc finger protein F7175; ZNF739; ZNF98 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 148198 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A6NK75 | |
| ZNF98 | |
| KLH conjugated synthetic peptide between 520-549 amino acids from the C-terminal region of human ZNF98. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction