Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ZO-1 Rat anti-Mouse, Rat, Clone: R40.76

Catalog No. MABT11MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT11MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT11MI Supplier MilliporeSigma™ Supplier No. MABT11
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-ZO-1 Antibody, clone R40.76 detects level of ZO-1 and has been published and validated for use in western blot, immunoprecipitation, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZO-1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone R40.76
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_033412
Gene Symbols ZO-1, ZO1
Host Species Rat
Immunogen Purified protein corresponding to mouse ZO-1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.