Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZRP-1 Mouse, Unlabeled, Clone: 16, BD

Catalog No. BDB612254 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612254 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612254 Supplier BD Biosciences Supplier No. 612254

Mouse Monoclonal Antibody

Zyxin is a zinc-binding metalloprotein found in focal adhesion plaques. It contains a proline-rich N-terminal domain and a C-terminal domain containing three LIM domains. LIM domains are characterized as zinc-binding motifs involved in gene expression and protein-protein interactions. Zyxin related protein 1 (ZRP-1) was identified through its interaction with the second PDZ domain of PTP1E. ZRP-1 has also been identified as Thyroid Receptor Interacting Protein-6 (TRIP6) during screening for binding partners of the thyroid hormone receptor.
ZRP-1 contains an N-terminal proline-rich region (PRR), and three double zinc finger LIM domains in the C-terminal region. ZRP-1 mRNA has been reported to be widely expressed with strong expression in heart, placenta, lung, liver, kidney, and pancreas. ZRP-1 may interact with focal adhesions similar to other LIM domain proteins in the zyxin-related subfamily. However, ZRP-1 also contains nuclear export signal, and treatment of cells with leptomycin B leads to movement of ZRP-1 from the cytoplasm to the nucleus. Thus, ZRP-1 may be important for relaying signals from focal adhesions to the nucleus.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZRP-1
Applications Western Blot
Classification Monoclonal
Clone 16
Concentration 250μg/mL
Conjugate Unconjugated
Description Zyxin Related Protein-1; TRIP6; Thyroid Receptor Interacting Protein-6
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ZRP-1 aa. 103-209
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.