Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zyxin Mouse anti-Human, Rat, Mouse, Clone: 14E11.1, MilliporeSigma™

Catalog No. MAB2610MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2610MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2610MI Supplier MilliporeSigma Supplier No. MAB2610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Zyxin Antibody, clone 14E11.1 is validated for use in western blot, IC for the detection of Zyxin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zyxin
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 14E11.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_001010972
Gene Symbols ESP-2, HED-2, ZYX
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human Zyxin.
Purification Method Ion Exchange
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.